Diethanolamine hydrochloride ≥95.0% (by total nitrogen and titration analysis)

Supplier: TCI America

Synonyms: 2,2'-Iminodiethanol Hydrochloride, 2,2'-Dihydroxydiethylamine Hydrochloride

I0009-25G I0009-500G
TCI0009-025GEA 41.08 CAD
TCI0009-025G TCI0009-500G
Diethanolamine hydrochloride ≥95.0% (by total nitrogen and titration analysis)
Diethanolamine hydrochloride

CAS Number: 14426-21-2
MDL Number: MFCD00012595
Molecular Formula: C4H11NO2
Molecular Weight: 141.60
Purity/Analysis Method: >95.0% (T,N)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.24


Other Product Names / Synonyms: 2,2'-Dihydroxydiethylamine Hydrochloride, 2,2'-Iminodiethanol Hydrochloride

Formula: C₄H₁₁NO₂
MW: 141.60 g/mol
Density: 1.24 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00012595
CAS Number: 14426-21-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5fadb5e63ec34fbe8761cb4f047aedb131c4c3f&vid=c5fadb5e63ec34fbe8761cb4f047aedb131c4c3f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR