Iminodiacetic acid disodium salt ≥98.0% for synthesis

Supplier: TCI America

Synonyms: Sodium 2,2'-azanediyldiacetate, Iminodiacetic acid disodium salt, Disodium Iminodiacetate hydrate

I0007-25G I0007-250G
TCI0007-025GEA 92.6 CAD
TCI0007-025G TCI0007-250G
Iminodiacetic acid disodium salt ≥98.0% for synthesis
Iminodiacetic acid disodium salt

CAS Number: 928-72-3
MDL Number: MFCD00043340
Molecular Formula: C4H7NO4
Molecular Weight: 177.07
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: Iminodiacetic Acid Disodium Salt

Formula: C₄H₅NNa₂O₄
MW: 177.07 g/mol
MDL Number: MFCD00043340
CAS Number: 928-72-3
EINECS: 213-181-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70e43a0a2293f5c66ca38586cc75d207e3679214&vid=70e43a0a2293f5c66ca38586cc75d207e3679214&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR