9-(2-Hydroxyethyl)adenine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
H1290-5G
H1290-25G
TCH1290-5GEA
90.06
CAD
TCH1290-5G
TCH1290-25G
9-(2-Hydroxyethyl)adenine ≥98.0% (by HPLC, titration analysis)
9-(2-Hydroxyethyl)adenine
CAS Number: 707-99-3
MDL Number: MFCD04220770
Molecular Formula: C7H9N5O
Molecular Weight: 179.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 242
Lambda max.: 261 nm (H2O)
Product Categories: Bioscience, Nucleobases and Their Analogs, Nucleosides and Their Analogs
Other Product Names / Synonyms: 6-Amino-9-(2-hydroxyethyl)purine
Formula:
C₇H₉N₅O Melting Pt: 242 °C |
MDL Number:
MFCD04220770 CAS Number: 707-99-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...