N-Hexadecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-16, SB-16) ≥98.0% (by total nitrogen basis) for biochemical research

Supplier: TCI America

Synonyms: Palmityldimethyl(3-sulphopropyl)ammonium Hydroxide Inner salt, Hexadecyldimethyl(3-sulphopropyl)ammonium Hydroxide Inner salt, 3-[Dimethyl(palmityl)ammonio]propane-1-sulfonate, Palmityl sulfobetaine, 3-[Dimethyl(palmityl)ammonio]propane-1-sulphonate, Palmityldimethyl(3-sulfopropyl)ammonium Hydroxide Inner salt, Hexadecyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner salt, Palmityl sulphobetaine, 3-(Hexadecyldimethylammonio)propane-1-sulphonate, 3-(Hexadecyldimethylammonio)propane-1-sulfonate

H1283-25G H1283-5G
TCH1283-25GEA 414.8 CAD
TCH1283-25G TCH1283-5G
N-Hexadecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-16, SB-16) ≥98.0% (by total nitrogen basis) for biochemical research
N-Hexadecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate (Sulfobetaine-16, SB-16)

[for Biochemical Research]
CAS Number: 2281-11-0
MDL Number: MFCD00036911
Molecular Formula: C21H45NO3S
Molecular Weight: 391.66
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 242
Flash Point (°C): 110


Product Categories: Bioscience, Amphoteric Surfactants (for Biochemistry)
Other Product Names / Synonyms: 3-(Hexadecyldimethylammonio)propane-1-sulfonate, Palmityldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt, Palmityl Sulfobetaine, 3-[Dimethyl(palmityl)ammonio]propane-1-sulfonate

Formula: C₂₁H₄₅NO₃S
MW: 391.6500 g/mol
Melting Pt: 242 °C
Flash Pt: 110 °C
MDL Number: MFCD00036911
CAS Number: 2281-11-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e95b2d8447659b3709f61fe5e5c1022342c9a886&vid=56ddec18cd059e136d1df0fce2515e11a39b873c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR