Hexyl methanesulfonate ≥96.0%

Supplier: TCI America

Synonyms: Methanesulphonic Acid Hexyl Ester, Methanesulfonic Acid Hexyl Ester

H1282-5G H1282-25G
TCH1282-5GEA 71.1 CAD
TCH1282-5G TCH1282-25G
Hexyl methanesulfonate ≥96.0%
Hexyl methanesulfonate

CAS Number: 16156-50-6
MDL Number: MFCD00674707
Molecular Formula: C7H16O3S
Molecular Weight: 180.26
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Specific Gravity (20/20): 1.06


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: Methanesulfonic Acid Hexyl Ester

Formula: C₇H₁₆O₃S
Boiling Pt: 90 °C
Density: 1.06 (20 °C)
MDL Number: MFCD00674707
CAS Number: 16156-50-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bcde0b414e6a075a9325237ec65a9cdad7f8e12&vid=0bcde0b414e6a075a9325237ec65a9cdad7f8e12&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR