Hexafluoro-2,3-bis(trifluoromethyl)-2,3-butanediol ≥98.0%

Supplier: TCI America

H1279-25G H1279-5G
TCH1279-25GEA 565.64 CAD
TCH1279-25G TCH1279-5G
Hexafluoro-2,3-bis(trifluoromethyl)-2,3-butanediol ≥98.0%
Hexafluoro-2,3-bis(trifluoromethyl)-2,3-butanediol

CAS Number: 918-21-8
MDL Number: MFCD00427700
Molecular Formula: C6H2F12O2
Molecular Weight: 334.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 129
Melting point (°C): 26


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks, Materials Chemistry, Monomers
Other Product Names / Synonyms: Perfluoropinacol

Formula: C₆H₂F₁₂O₂
Boiling Pt: 129 °C
Melting Pt: 26 °C
MDL Number: MFCD00427700
CAS Number: 918-21-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1741ad6d11f01558455fb10ad7a1bb322c70813e&vid=1741ad6d11f01558455fb10ad7a1bb322c70813e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR