4-Amino-6-hydroxy-2-methylpyrimidine hydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
['6-Amino-2-methyl-4-pyrimidinol Hydrate', '6-Amino-2-methyl-3', 'H', '-pyrimidin-4-one Hydrate'], 4-Amino-6-hydroxy-2-methylpyrimidine Hydrate, 6-Amino-2-methyl-3H-pyrimidin-4-one hydrate
A1470-5G
A1470-25G
TCA1470-005GEA
44.24
CAD
TCA1470-005G
TCA1470-025G
4-Amino-6-hydroxy-2-methylpyrimidine hydrate ≥98.0% (by titrimetric analysis)
4-Amino-6-hydroxy-2-methylpyrimidine hydrate
CAS Number: 767-16-8
MDL Number: MFCD00023266
Molecular Formula: C5H7N3O
Molecular Weight: 125.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: 6-Amino-2-methyl-4-pyrimidinol, 6-Amino-2-methyl-3H-pyrimidin-4-one
Formula:
C₅H₇N₃O MW: 143.14 g/mol |
MDL Number:
MFCD00150312 CAS Number: 732304-23-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...