DL-allo-Threonine ≥98.0% (contains Threonine) for synthesis

Supplier: TCI America

A0214-1G A0214-100MG
TCA0214-001GEA 496.12 CAD
TCA0214-001G TCA0214-100MG
DL-allo-Threonine ≥98.0% (contains Threonine) for synthesis
DL-allo-Threonine

CAS Number: 144-98-9
MDL Number: MFCD00067248
Molecular Formula: C4H9NO3
Molecular Weight: 119.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: DL-allo-Threonine

Formula: C₄H₉NO₃
MW: 119.12 g/mol
MDL Number: MFCD00067248
CAS Number: 144-98-9
EINECS: 205-645-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=08161595802220baf23cbb33528d546524b8d070&vid=08161595802220baf23cbb33528d546524b8d070&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR