8-Hydroxyquinoline-N-Oxide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 8-Quinolinol N-Oxide, 8-Hydroxyquinoline 1-oxide

H1272-5G H1272-25G
TCH1272-5GEA 71.1 CAD
TCH1272-5G TCH1272-25G
8-Hydroxyquinoline-N-Oxide ≥98.0% (by HPLC, titration analysis)
8-Hydroxyquinoline-N-Oxide

CAS Number: 1127-45-3
MDL Number: MFCD00006739
Molecular Formula: C9H7NO2
Molecular Weight: 161.16
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 139


Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: 8-Quinolinol N-Oxide

Formula: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: 139 °C
MDL Number: MFCD00006739
CAS Number: 1127-45-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e75f5f80cb6312d4aecb2a9d21eaaceba8225966&vid=e75f5f80cb6312d4aecb2a9d21eaaceba8225966&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR