2-[2-Hydroxy-5-[2-(methacryloyloxy)ethyl]phenyl]-2H-benzotriazole ≥98.0%

Supplier: TCI America

H1261-500G H1261-25G
TCH1261-500GEA 513.5 CAD
TCH1261-500G TCH1261-25G
2-[2-Hydroxy-5-[2-(methacryloyloxy)ethyl]phenyl]-2H-benzotriazole ≥98.0%
2-[2-Hydroxy-5-[2-(methacryloyloxy)ethyl]phenyl]-2H-benzotriazole

CAS Number: 96478-09-0
MDL Number: MFCD00213348
Molecular Formula: C18H17N3O3
Molecular Weight: 323.35
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 98

Formula: C₁₈H₁₇N₃O₃
MW: 323.35 g/mol
Melting Pt: 98 °C
MDL Number: MFCD00213348
CAS Number: 96478-09-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c05d79f7feafc3ade12b8ad95458824986e94293&vid=c05d79f7feafc3ade12b8ad95458824986e94293&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR