trans-4-(Hydroxymethyl)cyclohexanol ≥98.0%
Supplier: TCI America
H1258-1G
H1258-5G
TCH1258-1GEA
476.05
CAD
TCH1258-1G
TCH1258-5G
trans-4-(Hydroxymethyl)cyclohexanol ≥98.0%
trans-4-(Hydroxymethyl)cyclohexanol
CAS Number: 3685-27-6
Molecular Formula: C7H14O2
Molecular Weight: 130.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 104
Formula:
C₇H₁₄O₂ Melting Pt: 104 °C |
MDL Number:
MFCD11036248 CAS Number: 3685-27-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...