4-(Hydroxymethyl)cyclohexanol (cis- and trans- mixture) ≥98.0%
Supplier: TCI America
H1257-25G
H1257-5G
TCH1257-25GEA
1504.16
CAD
TCH1257-25G
TCH1257-5G
4-(Hydroxymethyl)cyclohexanol (cis- and trans- mixture) ≥98.0%
4-(Hydroxymethyl)cyclohexanol (cis- and trans- mixture)
CAS Number: 33893-85-5
Molecular Formula: C7H14O2
Molecular Weight: 130.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 122
Formula:
C₇H₁₄O₂ Boiling Pt: 122 °C |
MDL Number:
MFCD11036248 CAS Number: 33893-85-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...