4-(2-Hydroxyethyl)cyclohexanol (cis- and trans- mixture) ≥98.0%

Supplier: TCI America

Synonyms: 2-(4-Hydroxycyclohexyl)ethanol, 4-Hydroxycyclohexaneethanol

H1253-5G H1253-1G
TCH1253-5GEA 618.25 CAD
TCH1253-5G TCH1253-1G
4-(2-Hydroxyethyl)cyclohexanol (cis- and trans- mixture) ≥98.0%
4-(2-Hydroxyethyl)cyclohexanol (cis- and trans- mixture)

CAS Number: 74058-21-2
Molecular Formula: C8H16O2
Molecular Weight: 144.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 175
Specific Gravity (20/20): 1.05


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 2-(4-Hydroxycyclohexyl)ethanol, 4-Hydroxycyclohexaneethanol

Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 175 °C
Density: 1.05 (20 °C)
CAS Number: 74058-21-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74ed1da14cf792a3b4e89dfeffb19c1aa3cdef15&vid=74ed1da14cf792a3b4e89dfeffb19c1aa3cdef15&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR