trans-4-(Hydroxymethyl)cyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
H1243-5G
H1243-1G
TCH1243-5GEA
593.45
CAD
TCH1243-5G
TCH1243-1G
trans-4-(Hydroxymethyl)cyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
trans-4-(Hydroxymethyl)cyclohexanecarboxylic acid
CAS Number: 66185-74-8
MDL Number: MFCD03840884
Molecular Formula: C8H14O3
Molecular Weight: 158.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 141
Formula:
C₈H₁₄O₃ MW: 158.20 g/mol Melting Pt: 141 °C |
MDL Number:
MFCD03840884 CAS Number: 66185-74-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...