cis-4-(Hydroxymethyl)cyclohexanecarboxylic acid ≥98.0%

Supplier: TCI America

H1242-5G H1242-25G
TCH1242-5GEA 294.05 CAD
TCH1242-5G TCH1242-25G
cis-4-(Hydroxymethyl)cyclohexanecarboxylic acid ≥98.0%
cis-4-(Hydroxymethyl)cyclohexanecarboxylic acid

CAS Number: 73094-35-6
Molecular Formula: C8H14O3
Molecular Weight: 158.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 120

Formula: C₈H₁₄O₃
MW: 158.20 g/mol
Melting Pt: 120 °C
CAS Number: 73094-35-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0acd398e9a7d83a96d4fed8539f76ec6f81700b4&vid=0acd398e9a7d83a96d4fed8539f76ec6f81700b4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR