D-(+)-Homoserine 96,0

Supplier: TCI America

Synonyms: H-D-Hser-OH, H-D-HomoSer-OH, H-D-Homoser-OH, (R)-(+)-Homoserine, (R)-2-Amino-4-hydroxybutyric acid, D-Homoserine

H1224-1G H1224-5G
TCH1224-1GEA 137.46 CAD
TCH1224-1G TCH1224-5G
D-(+)-Homoserine 96,0
D(+)-Homoserine

CAS Number: 6027-21-0
MDL Number: MFCD00077786
Molecular Formula: C4H9NO3
Molecular Weight: 119.12
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Melting point (°C): 204
Specific rotation [a]20/D: 8 deg (C=2, H2O)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (R)-2-Amino-4-hydroxybutyric Acid

Formula: C₄H₉NO₃
MW: 119.12
Melting Pt: 204 °C
MDL Number: MFCD00077786
CAS Number: 6027-21-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8d1eedd72c113177fb8edd18f27b64696915a27&vid=c8d1eedd72c113177fb8edd18f27b64696915a27&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR