4-(11-Hydroxyundecyloxy)benzaldehyde ≥98.0% (by HPLC)

Supplier: TCI America

H1216-1G H1216-5G
TCH1216-1GEA 48.98 CAD
TCH1216-1G TCH1216-5G
4-(11-Hydroxyundecyloxy)benzaldehyde ≥98.0% (by HPLC)
4-(11-Hydroxyundecyloxy)benzaldehyde

CAS Number: 124389-14-6
Molecular Formula: C18H28O3
Molecular Weight: 292.42
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 64

Formula: C₁₈H₂₈O₃
MW: 292.42 g/mol
Melting Pt: 64 °C
CAS Number: 124389-14-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3ff0aa5aac41a9be3cba361ae78ed58a56ea592&vid=f3ff0aa5aac41a9be3cba361ae78ed58a56ea592&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR