D-Histidine methyl ester dihydrochloride ≥97.0% (by total nitrogen basis)

Supplier: TCI America

H1213-1G H1213-5G
TCH1213-1GEA 42.66 CAD
TCH1213-1G TCH1213-5G
D-Histidine methyl ester dihydrochloride ≥97.0% (by total nitrogen basis)
D-Histidine methyl ester dihydrochloride

CAS Number: 4467-54-3
MDL Number: MFCD00066137
Molecular Formula: C7H11N3O2
Molecular Weight: 242.10
Purity/Analysis Method: >97.0% (N)
Form: Crystal
Melting point (°C): 197
Specific rotation [a]20/D: -9 deg (C=2, H2O)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-D-His-OMe.2HCl

Formula: C₇H₁₁N₃O₂
Melting Pt: 197 °C
MDL Number: MFCD00066137
CAS Number: 4467-54-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58ef01594d5d609d54380182f645fb3e7df71a10&vid=58ef01594d5d609d54380182f645fb3e7df71a10&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR