3',4'-Dimethoxy-2'-hydroxyacetophenone ≥98.0%

Supplier: TCI America

H1205-5G H1205-25G
TCH1205-5GEA 75.84 CAD
TCH1205-5G TCH1205-25G
3',4'-Dimethoxy-2'-hydroxyacetophenone ≥98.0%
3',4'-Dimethoxy-2'-hydroxyacetophenone

CAS Number: 5396-18-9
MDL Number: MFCD00016684
Molecular Formula: C10H12O4
Molecular Weight: 196.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 77

Formula: C₁₀H₁₂O₄
Melting Pt: 77 °C
MDL Number: MFCD00016684
CAS Number: 5396-18-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=479940c62ab16e8d6670642a8b9e66b21aaa06e9&vid=479940c62ab16e8d6670642a8b9e66b21aaa06e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR