2,3,6,7,10,11-Hexabromotriphenylene ≥98.0% (by titrimetric analysis)

Supplier: TCI America

H1201-100MG H1201-1G
TCH1201-100MGEA 75.84 CAD
TCH1201-100MG TCH1201-1G
2,3,6,7,10,11-Hexabromotriphenylene ≥98.0% (by titrimetric analysis)
2,3,6,7,10,11-Hexabromotriphenylene

CAS Number: 82632-80-2
Molecular Formula: C18H6Br6
Molecular Weight: 701.67
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₁₈H₆Br₆
MW: 701.67 g/mol
MDL Number: MFCD09834716
CAS Number: 82632-80-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=140ed6256d80d25ff5f211479985e2629634a0db&vid=140ed6256d80d25ff5f211479985e2629634a0db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR