1,2,3,4,8,9,10,11,15,16,17,18,22,23,24,25-Hexadecafluorophthalocyaninecopper(II) ≥98.0% (by total nitrogen and titration analysis), purified by sublimation

Supplier: TCI America

H1194-1G H1194-100MG
TCH1194-1GEA 379.2 CAD
TCH1194-1G TCH1194-100MG
1,2,3,4,8,9,10,11,15,16,17,18,22,23,24,25-Hexadecafluorophthalocyaninecopper(II) ≥98.0% (by total nitrogen and titration analysis), purified by sublimation
1,2,3,4,8,9,10,11,15,16,17,18,22,23,24,25-Hexadecafluorophthalocyaninecopper(II)

CAS Number: 14916-87-1
MDL Number: MFCD00274642
Molecular Formula: C32CuF16N8
Molecular Weight: 863.93
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal


Product Categories: Materials Chemistry, Organic Transistor Materials, Solar Cell Research Reagents, Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Highly Purified Reagents, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: Copper(II) 1,2,3,4,8,9,10,11,15,16,17,18,22,23,24,25-Hexadecafluorophthalocyanine, F16CuPc

Formula: C₃₂CuF₁₆N₈
MDL Number: MFCD00274642
CAS Number: 14916-87-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bb6ffc149a0ac972d8953a21078e60c823a54cc&vid=9dc83f202c6f0f5f49f7dced5cd1ca7dc004a362&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR