2-(2-(Piperazin-1-yl)ethoxy)ethanol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
1-[2-(2-Hydroxyethoxy)ethyl]piperazine, 1-Hydroxyethylethoxypiperazine
H1192-5G
H1192-25G
TCH1192-5GEA
58.46
CAD
TCH1192-5G
TCH1192-25G
2-(2-(Piperazin-1-yl)ethoxy)ethanol ≥98.0% (by GC, titration analysis)
2-(2-(Piperazin-1-yl)ethoxy)ethanol
CAS Number: 13349-82-1
MDL Number: MFCD00074931
Molecular Formula: C8H18N2O2
Molecular Weight: 174.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 172
Flash Point (°C): 113
Specific Gravity (20/20): 1.08
Other Product Names / Synonyms: 2-[2-(1-Piperazinyl)ethoxy]ethanol
Formula:
C₈H₁₈N₂O₂ MW: 174.2407 g/mol Boiling Pt: 172 °C Density: 1.08 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00074931 CAS Number: 13349-82-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...