trans-4-Hydroxycyclohexanecarboxylic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

H1175-25G H1175-5G
TCH1175-25GEA 325.48 CAD
TCH1175-25G TCH1175-5G
trans-4-Hydroxycyclohexanecarboxylic acid ≥97.0% (by GC, titration analysis)
trans-4-Hydroxycyclohexanecarboxylic acid

CAS Number: 3685-26-5
Molecular Formula: C7H12O3
Molecular Weight: 144.17
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 145

Formula: C₇H₁₂O₃
MW: 144.1684 g/mol
Melting Pt: 145 °C
MDL Number: MFCD02093477
CAS Number: 3685-26-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b92eb5e3224bc5fa5668397bd1e693ca48584b3&vid=5b92eb5e3224bc5fa5668397bd1e693ca48584b3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR