2-Hydroxy-3-nitro-4-picoline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
H1170-1G
H1170-5G
TCH1170-1GEA
64.78
CAD
TCH1170-1G
TCH1170-5G
2-Hydroxy-3-nitro-4-picoline ≥98.0% (by GC, titration analysis)
2-Hydroxy-3-nitro-4-picoline
CAS Number: 21901-18-8
Molecular Formula: C6H6N2O3
Molecular Weight: 154.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 234
Other Product Names / Synonyms: 2-Hydroxy-3-nitro-4-picoline, 4-Methyl-3-nitro-2-pyridinol, 4-Methyl-3-nitro-2-pyridone
Formula:
C₆H₆N₂O₃ Melting Pt: 234 °C |
MDL Number:
MFCD00010689 CAS Number: 21901-18-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...