Hexadecyl acrylate 90,0 Stabilised

Supplier: TCI America

Synonyms: Acrylic Acid Hexadecyl Ester

H1168-25G H1168-250G
TCH1168-25GEA 117.3 CAD
TCH1168-25G TCH1168-250G
Hexadecyl acrylate 90,0 Stabilised
Hexadecyl acrylate

(stabilized with MEHQ)
CAS Number: 13402-02-3
Molecular Formula: C19H36O2
Molecular Weight: 296.50
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Flash Point (°C): 178
Freezing point (°C): 17
Specific Gravity (20/20): 0.87


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Acrylic Acid Hexadecyl Ester

Formula: C₁₉H₃₆O₂
MW: 296.50 g/mol
Density: 0.87 (20 °C)
Flash Pt: 178 °C
MDL Number: MFCD00078441
CAS Number: 13402-02-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e80adb5fee7151baab7459301677dcb7edf69544&vid=e80adb5fee7151baab7459301677dcb7edf69544&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR