3-Amino-5-methylthio-1H-1,2,4-triazole ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3-Amino-5-(methylmercapto)-1H-1,2,4-triazole

A1462-25G A1462-5G
TCA1462-025GEA 148.52 CAD
TCA1462-025G TCA1462-5G
3-Amino-5-methylthio-1H-1,2,4-triazole ≥97.0% (by titrimetric analysis)
3-Amino-5-methylthio-1H-1,2,4-triazole

CAS Number: 45534-08-5
MDL Number: MFCD00005232
Molecular Formula: C3H6N4S
Molecular Weight: 130.17
Purity/Analysis Method: >97.0% (T)
Form: Crystal

Formula: C₃H₆N₄S
MW: 130.17 g/mol
MDL Number: MFCD00005232
CAS Number: 45534-08-5
EINECS: 256-242-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7c4702aef0b3ce5613054bbbadecfd3b16c12b14&vid=7c4702aef0b3ce5613054bbbadecfd3b16c12b14&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR