Hexadecyl lactate ≥75.0%

Supplier: TCI America

Synonyms: Lactic Acid Hexadecyl Ester, Hexadecyl 2-Hydroxypropionate, Cetyl Lactate

H1159-25G H1159-500G
TCH1159-25GEA 47.4 CAD
TCH1159-25G TCH1159-500G
Hexadecyl lactate ≥75.0%
Hexadecyl lactate

CAS Number: 35274-05-6
Molecular Formula: C19H38O3
Molecular Weight: 314.51
Purity/Analysis Method: >75.0% (GC)
Form: Crystal
Melting point (°C): 42


Other Product Names / Synonyms: Cetyl Lactate, Hexadecyl 2-Hydroxypropionate, Lactic Acid Hexadecyl Ester

Formula: C₁₉H₃₈O₃
Melting Pt: 42 °C
CAS Number: 35274-05-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb99b4cef851323e3725d5ac0d2b2cc1a13d429f&vid=cb99b4cef851323e3725d5ac0d2b2cc1a13d429f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 75.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR