Hexyl-7-(diethylamino)coumarin-3-carboxylate ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 7-(Diethylamino)coumarin-3-carboxylic Acid Hexyl Ester

H1145-5G H1145-1G
TCH1145-5GEA 404.95 CAD
TCH1145-5G TCH1145-1G
Hexyl-7-(diethylamino)coumarin-3-carboxylate ≥98.0% (by HPLC)
Hexyl-7-(diethylamino)coumarin-3-carboxylate

CAS Number: 851963-03-6
MDL Number: MFCD06797113
Molecular Formula: C20H27NO4
Molecular Weight: 345.44
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Product Categories: Materials Chemistry, Coumarin Dyes (Functional Organic Dyes), Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: 7-(Diethylamino)coumarin-3-carboxylic Acid Hexyl Ester

Formula: C₂₀H₂₇NO₄
MW: 345.44 g/mol
MDL Number: MFCD06797113
CAS Number: 851963-03-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7124985826163b0c7457d7c3300c60408864f31c&vid=7124985826163b0c7457d7c3300c60408864f31c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR