4-(3-Hydroxypropyl)thiomorpholine-1,1-dioxide ≥98.0%

Supplier: TCI America

H1141-5G H1141-25G
TCH1141-5GEA 266.07 CAD
TCH1141-5G TCH1141-25G
4-(3-Hydroxypropyl)thiomorpholine-1,1-dioxide ≥98.0%
4-(3-Hydroxypropyl)thiomorpholine-1,1-dioxide

CAS Number: 205194-33-8
Molecular Formula: C7H15NO3S
Molecular Weight: 193.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 72


Other Product Names / Synonyms: 3-(1,1-Dioxothiomorpholino)-1-propanol

Formula: C₇H₁₅NO₃S
Melting Pt: 72 °C
CAS Number: 205194-33-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c277adb33b33c8fd064d670b7c667fbaae5b9f97&vid=c277adb33b33c8fd064d670b7c667fbaae5b9f97&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR