4-(2-Hydroxyethyl)thiomorpholine-1,1-dioxide ≥98.0%

Supplier: TCI America

Synonyms: 4-(2-Hydroxyethyl)thiomorpholine 1,1-Dioxide, 2-(1,1-Dioxothiomorpholino)ethanol

H1140-25G H1140-5G
TCH1140-25GEA 489.33 CAD
TCH1140-25G TCH1140-5G
4-(2-Hydroxyethyl)thiomorpholine-1,1-dioxide ≥98.0%
4-(2-Hydroxyethyl)thiomorpholine-1,1-dioxide

CAS Number: 26475-62-7
Molecular Formula: C6H13NO3S
Molecular Weight: 179.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 197
Melting point (°C): 74


Other Product Names / Synonyms: 2-(1,1-Dioxothiomorpholino)ethanol

Formula: C₆H₁₃NO₃S
MW: 179.23 g/mol
Boiling Pt: 197 °C
Melting Pt: 74 °C
MDL Number: MFCD00106415
CAS Number: 26475-62-7

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR