2-Amino-5,6-dimethylbenzothiazole ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
5,6-Dimethylbenzo[d]thiazol-2-amine
A1461-5G
A1461-25G
TCA1461-005GEA
371.3
CAD
TCA1461-005G
TCA1461-025G
2-Amino-5,6-dimethylbenzothiazole ≥98.0% (by HPLC, titration analysis)
2-Amino-5,6-dimethylbenzothiazole
CAS Number: 29927-08-0
MDL Number: MFCD00005790
Molecular Formula: C9H10N2S
Molecular Weight: 178.25
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 190
Formula:
C₉H₁₀N₂S MW: 178.2500 g/mol Melting Pt: 190 °C |
MDL Number:
MFCD00005790 CAS Number: 29927-08-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...