5-Hydroxy-1-indanone ≥98.0%
Supplier: TCI America
H1051-5G
H1051-25G
TCH1051-5GEA
134.3
CAD
TCH1051-5G
TCH1051-25G
5-Hydroxy-1-indanone ≥98.0%
5-Hydroxy-1-indanone
CAS Number: 3470-49-3
MDL Number: MFCD00857527
Molecular Formula: C9H8O2
Molecular Weight: 148.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 184
Formula:
C9H8O2 Melting Pt: 184 °C |
MDL Number:
MFCD00857527 CAS Number: 3470-49-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...