m-PEG7-alcohol ≥97.0%

Supplier: TCI America

H1046-1G H1046-5G
TCH1046-1GEA 178.54 CAD
TCH1046-1G TCH1046-5G
m-PEG7-alcohol ≥97.0%
m-PEG7-alcohol

CAS Number: 4437-01-8
MDL Number: MFCD06201006
Molecular Formula: C15H32O8
Molecular Weight: 340.41
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.09


Product Categories: Bioscience, PEGylation Reagents
Other Product Names / Synonyms: mPEG7-Alcohol

Formula: C₁₅H₃₂O₈
Density: 1.09 (20 °C)
MDL Number: MFCD06201006
CAS Number: 4437-01-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=803e99222a90ee05cd7172dbae74cacab975a2f1&vid=803e99222a90ee05cd7172dbae74cacab975a2f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR