Ethyl (R)-(-)-3-hydroxybutyrate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl (R)-3-hydroxybutyrate, (R)-(-)-3-Hydroxybutyric Acid Ethyl Ester

H1029-1G H1029-5G
TCH1029-1GEA 102.7 CAD
TCH1029-1G TCH1029-5G
Ethyl (R)-(-)-3-hydroxybutyrate ≥98.0%
Ethyl (R)-(-)-3-hydroxybutyrate

CAS Number: 24915-95-5
MDL Number: MFCD00075386
Molecular Formula: C6H12O3
Molecular Weight: 132.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 64
Specific rotation [a]20/D: -18 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, Lipids
Other Product Names / Synonyms: (R)-(-)-3-Hydroxybutyric Acid Ethyl Ester

Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Flash Pt: 64 °C
MDL Number: MFCD00075386
CAS Number: 24915-95-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb1b53b37d3e8bacf31945af8e548b06d00e47d0&vid=fb1b53b37d3e8bacf31945af8e548b06d00e47d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR