(S)-(+)-O-Acetylmandelic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A1454-25G A1454-5G
TCA1454-25GEA 287.56 CAD
TCA1454-25G TCA1454-005G
(S)-(+)-O-Acetylmandelic acid ≥98.0% (by titrimetric analysis)
(S)-(+)-O-Acetylmandelic acid

CAS Number: 7322-88-5
MDL Number: MFCD00064215
Molecular Formula: C10H10O4
Molecular Weight: 194.19
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 98
Specific rotation [a]20/D: 153 deg (C=2, Acetone)


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Optical Resolving Agents
Other Product Names / Synonyms: (S)-(+)-alpha-Acetoxyphenylacetic Acid

Formula: C₁₀H₁₀O₄
Melting Pt: 98 °C
MDL Number: MFCD00064215
CAS Number: 7322-88-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6365b0e03e0df1105f559bf5a37b42883809a3f4&vid=6365b0e03e0df1105f559bf5a37b42883809a3f4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR