4-Hydroxyphenyldimethylsulfonium methyl sulfate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
H1022-25G
H1022-5G
TCH1022-25GEA
369.72
CAD
TCH1022-25G
TCH1022-5G
4-Hydroxyphenyldimethylsulfonium methyl sulfate ≥98.0% (by titrimetric analysis)
4-Hydroxyphenyldimethylsulfonium methyl sulfate
CAS Number: 32279-04-2
Molecular Formula: C9H14O5S2
Molecular Weight: 266.33
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 92
Other Product Names / Synonyms: Dimethyl-4-hydroxyphenylsulfonium Methyl Sulfate
Formula:
C₉H₁₄O₅S₂ Melting Pt: 92 °C |
MDL Number:
MFCD08276380 CAS Number: 32279-04-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...