3-Hydroxy-2,6-bis(hydroxymethyl)-4-pyrone ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
H1019-5G
H1019-1G
TCH1019-5GEA
417.12
CAD
TCH1019-5G
TCH1019-1G
3-Hydroxy-2,6-bis(hydroxymethyl)-4-pyrone ≥98.0% (by HPLC, titration analysis)
3-Hydroxy-2,6-bis(hydroxymethyl)-4-pyrone
CAS Number: 2029-49-4
MDL Number: MFCD00110732
Molecular Formula: C7H8O5
Molecular Weight: 172.14
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 149
Formula:
C₇H₈O₅ Melting Pt: 149 °C |
MDL Number:
MFCD00110732 CAS Number: 2029-49-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...