4-(Hydroxymethyl)cyclohexanecarboxylic acid (cis and trans mixture) ≥98.0%
Supplier: TCI America
Synonyms:
4-(Hydroxymethyl)cyclohexanecarboxylic acid, 4-Carboxy-1-cyclohexanemethanol
H1015-5G
H1015-25G
TCH1015-5GEA
270.18
CAD
TCH1015-5G
TCH1015-25G
4-(Hydroxymethyl)cyclohexanecarboxylic acid (cis and trans mixture) ≥98.0%
4-(Hydroxymethyl)cyclohexanecarboxylic acid (cis and trans mixture)
CAS Number: 13380-84-2
MDL Number: MFCD01734677
Molecular Formula: C8H14O3
Molecular Weight: 158.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Other Product Names / Synonyms: 4-Carboxy-1-cyclohexanemethanol
Formula:
C₈H₁₄O₃ |
MDL Number:
MFCD01734677 CAS Number: 13380-84-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...