1,1,2,2,3,3,4-Heptafluorocyclopentane ≥98.0%

Supplier: TCI America

H1013-25G H1013-500G
TCH1013-25GEA 97.49 CAD
TCH1013-25G TCH1013-500G
1,1,2,2,3,3,4-Heptafluorocyclopentane ≥98.0%
1,1,2,2,3,3,4-Heptafluorocyclopentane

CAS Number: 15290-77-4
MDL Number: MFCD03844796
Molecular Formula: C5H3F7
Molecular Weight: 196.07
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 83
Freezing point (°C): 21

Formula: C₅H₃F₇
Boiling Pt: 83 °C
MDL Number: MFCD03844796
CAS Number: 15290-77-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d99438ec184ad341334b50fa5942e5894b755f8f&vid=d99438ec184ad341334b50fa5942e5894b755f8f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR