2-Hydroxy-5-methylbenzophenone ≥97.0%

Supplier: TCI America

H1008-25G H1008-5G
TCH1008-25GEA 414.91 CAD
TCH1008-25G TCH1008-5G
2-Hydroxy-5-methylbenzophenone ≥97.0%
2-Hydroxy-5-methylbenzophenone

CAS Number: 1470-57-1
MDL Number: MFCD00002379
Molecular Formula: C14H12O2
Molecular Weight: 212.25
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 85

Formula: C₁₄H₁₂O₂
Melting Pt: 85 °C
MDL Number: MFCD00002379
CAS Number: 1470-57-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=250af38f0d8e2d7f1c3de55d754b9b643f0b156f&vid=250af38f0d8e2d7f1c3de55d754b9b643f0b156f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR