(R)-2-Acetoxy-2-phenylacetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

A1453-5G A1453-25G
TCA1453-005GEA 64.78 CAD
TCA1453-005G TCA1453-25G
(R)-2-Acetoxy-2-phenylacetic acid ≥98.0% (by GC, titration analysis)
(R)-2-Acetoxy-2-phenylacetic acid

CAS Number: 51019-43-3
MDL Number: MFCD00004249
Molecular Formula: C10H10O4
Molecular Weight: 194.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 98
Specific rotation [a]20/D: -153 deg (C=2, Acetone)


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Optical Resolving Agents
Other Product Names / Synonyms: (R)-(-)-alpha-Acetoxyphenylacetic Acid

Formula: C10H10O4
Melting Pt: 98 °C
MDL Number: MFCD00004249
CAS Number: 51019-43-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae44293485f62ed812734d5a422e5bd530b40e02&vid=ae44293485f62ed812734d5a422e5bd530b40e02&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR