N-(Hydroxymethyl)nicotinamide ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

H1001-25G H1001-500G
TCH1001-25GEA 64.78 CAD
TCH1001-25G TCH1001-500G
N-(Hydroxymethyl)nicotinamide ≥97.0% (by HPLC, titration analysis)
N-(Hydroxymethyl)nicotinamide

CAS Number: 3569-99-1
MDL Number: MFCD00010437
Molecular Formula: C7H8N2O2
Molecular Weight: 152.15
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 145

Formula: C₇H₈N₂O₂
Melting Pt: 145 °C
MDL Number: MFCD00010437
CAS Number: 3569-99-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3d1b887518391e61def562a8f9297f2c555cb54&vid=f3d1b887518391e61def562a8f9297f2c555cb54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR