4(1H)-Pyrimidinone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3,4-Dihydropyrimidin-4-one, 1H-Pyrimidin-4-one, 4-Hydroxypyrimidine, Pyrimidin-4(1H)-one

H1000-5G H1000-1G
TCH1000-5GEA 137.46 CAD
TCH1000-5G TCH1000-1G
4(1H)-Pyrimidinone ≥98.0% (by GC, titration analysis)
4(1H)-Pyrimidinone

CAS Number: 4562-27-0
MDL Number: MFCD00167178
Molecular Formula: C4H4N2O
Molecular Weight: 96.09
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystalline Powder
Melting point (°C): 166


Other Product Names / Synonyms: 4-Hydroxypyrimidine, 4-Pyrimidinol

Formula: C₄H₄N₂O
MW: 96.09 g/mol
Melting Pt: 166 °C
MDL Number: MFCD00006664
CAS Number: 51953-17-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c5605e0ee214adf64c7b94d3628b76872ba53ac&vid=3a769ccb16f193acc6db736263fbeed5c561a373&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR