2-Hydroxy-2-methylpropiophenone ≥96.0%

Supplier: TCI America

H0991-25G H0991-500G H0991-100G
TCH0991-25GEA 95.04 CAD
TCH0991-25G TCH0991-500G TCH0991-100G
2-Hydroxy-2-methylpropiophenone ≥96.0%
2-Hydroxy-2-methylpropiophenone

CAS Number: 7473-98-5
MDL Number: MFCD00040710
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Flash Point (°C): 110
Specific Gravity (20/20): 1.08


Product Categories: Materials Chemistry, Polymerization Initiators
Other Product Names / Synonyms: 2-Benzoyl-2-propanol

Formula: C₁₀H₁₂O₂
Boiling Pt: 110 °C
Density: 1.08 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00040710
CAS Number: 7473-98-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e85ae67e8470380cfe86a69a7d05530785c1fed2&vid=e85ae67e8470380cfe86a69a7d05530785c1fed2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR