cis-4-Hydroxycyclohexanecarboxylic acid ≥98.0%

Supplier: TCI America

H0980-25G H0980-5G
TCH0980-25GEA 647.96 CAD
TCH0980-25G TCH0980-005G
cis-4-Hydroxycyclohexanecarboxylic acid ≥98.0%
cis-4-Hydroxycyclohexanecarboxylic acid

CAS Number: 3685-22-1
MDL Number: MFCD04038017
Molecular Formula: C7H12O3
Molecular Weight: 144.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 150

Formula: C₇H₁₂O₃
Melting Pt: 150 °C
MDL Number: MFCD02093477
CAS Number: 3685-22-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=054467cb2cff95346fbb2ff053416aef9d9068f4&vid=054467cb2cff95346fbb2ff053416aef9d9068f4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR