4-Hydroxymethyl-5-methylimidazole ≥96.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (5-Methyl-1H-imidazol-4-yl)methanol

H0976-25G H0976-5G
TCH0976-25GEA 246.48 CAD
TCH0976-25G TCH0976-5G
4-Hydroxymethyl-5-methylimidazole ≥96.0% (by GC, titration analysis)
4-Hydroxymethyl-5-methylimidazole

CAS Number: 29636-87-1
MDL Number: MFCD00792476
Molecular Formula: C5H8N2O
Molecular Weight: 112.13
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Melting point (°C): 136

Formula: C₅H₈N₂O
MW: 112.13 g/mol
Melting Pt: 136 °C
MDL Number: MFCD00792476
CAS Number: 29636-87-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d2d3206bbe316005b92fb5c732cd840e9076cb3&vid=0d2d3206bbe316005b92fb5c732cd840e9076cb3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR