(S)-(+)-Ethyl-3-hydroxybutyrate ≥96.0%

Supplier: TCI America

Synonyms: (S)-Ethyl-3-hydroxybutyrate, Ethyl (S)-3-hydroxybutanoate, Ethyl (S)-(+)-3-hydroxybutyrate, (S)-Ethyl 3-hydroxybutanoate, Ethyl (S)-3-hydroxybutyrate

H0975-25G H0975-5G
TCH0975-025GEA 740.55 CAD
TCH0975-025G TCH0975-005G
(S)-(+)-Ethyl-3-hydroxybutyrate ≥96.0%
(S)-(+)-Ethyl 3-hydroxybutyrate

CAS Number: 56816-01-4
MDL Number: MFCD00066206
Molecular Formula: C6H12O3
Molecular Weight: 132.16
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Flash Point (°C): 64
Specific Gravity (20/20): 1.01
Specific rotation [a]20/D: 18 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, Lipids
Other Product Names / Synonyms: (S)-(+)-3-Hydroxybutyric Acid Ethyl Ester

Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Density: 1.01 (20 °C)
Flash Pt: 64 °C
MDL Number: MFCD00066206
CAS Number: 56816-01-4
EINECS: 260-393-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26e1108352b069a131621b13105159c6c3bfdaa5&vid=26e1108352b069a131621b13105159c6c3bfdaa5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR