9-(2-Hydroxyethyl)anthracene ≥98.0%

Supplier: TCI America

H0970-5G H0970-1G
TCH0970-005GEA 1303.5 CAD
TCH0970-005G TCH0970-001G
9-(2-Hydroxyethyl)anthracene ≥98.0%
9-(2-Hydroxyethyl)anthracene

CAS Number: 54060-73-0
MDL Number: MFCD01318099
Molecular Formula: C16H14O
Molecular Weight: 222.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 125


Other Product Names / Synonyms: 2-(9-Anthryl)ethanol

Formula: C₁₆H₁₄O
Melting Pt: 125 °C
MDL Number: MFCD01318099
CAS Number: 54060-73-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=311d3ad9c0434684119f70e0347692242397cb07&vid=311d3ad9c0434684119f70e0347692242397cb07&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR