4-(3-Hydroxyphenyl)piperidine ≥98.0%

Supplier: TCI America

Synonyms: 3-(Piperidin-4-yl)phenol, 3-(4-Piperidyl)phenol

H0965-5G H0965-25G
TCH0965-005GEA 870.58 CAD
TCH0965-005G TCH0965-25G
4-(3-Hydroxyphenyl)piperidine ≥98.0%
4-(3-Hydroxyphenyl)piperidine

CAS Number: 110878-71-2
MDL Number: MFCD02093474
Molecular Formula: C11H15NO
Molecular Weight: 177.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 225


Other Product Names / Synonyms: 3-(4-Piperidyl)phenol

Formula: C₁₁H₁₅NO
MW: 177.25 g/mol
Melting Pt: 225 °C
MDL Number: MFCD02093474
CAS Number: 110878-71-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=990ed4d53307563241178d9d430e1e5416fdbe45&vid=990ed4d53307563241178d9d430e1e5416fdbe45&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR