(S)-(+)-2-Amino-4-bromobutyric acid hydrobromide ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America

Synonyms: (S)-2-Amino-4-bromobutanoic acid hydrobromide

A1450-1G A1450-5G
TCA1450-1GEA 135.04 CAD
TCA1450-1G TCA1450-005G
(S)-(+)-2-Amino-4-bromobutyric acid hydrobromide ≥98.0% (by total nitrogen and titration analysis)
(S)-(+)-2-Amino-4-bromobutyric acid hydrobromide

CAS Number: 15159-65-6
MDL Number: MFCD01631290
Molecular Formula: C4H8BrNO2
Molecular Weight: 262.93
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 184
Specific rotation [a]20/D: 17 deg (C=1, MeOH)

Formula: C₄H₈BrNO₂
MW: 262.93 g/mol
Melting Pt: 184 °C
MDL Number: MFCD01631290
CAS Number: 15159-65-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f19ff9c296c116220ac14486daefb2c99f76d85f&vid=f19ff9c296c116220ac14486daefb2c99f76d85f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR